Learn More
Cayman Chemical α-Phthalimidopropiophenone, Purity: ≥98%, CAS: 19437-20-8, 10 mg
SDP
Catalog No. 502967605
Encompass_Preferred
A cathinone that may act as a prodrug, forming an active cathinone following metabolism in vivo; intended for forensic and research applications
Catalog No. 50-296-7605 Supplier Cayman Chemical Supplier No. 1714110 MG
A cathinone that may act as a prodrug, forming an active cathinone following metabolism in vivo; intended for forensic and research applications