Learn More
Selleck Chemical LLC a-Pinene
SDP
Supplier:  Selleck Chemical LLC S559625UL
Encompass_Preferred
-Pinene is found in the oils of many species of many coniferous trees notably the pine and has insecticidal activity (1S)-(-)- -Pinene is a monoterpene and shows sleep enhancing property through a direct binding to GABAA-benzodiazepine (BZD) receptors by acting as a partial modulator at the BZD binding site
Catalog No. 50-000-03514