Learn More
Cayman Chemical α-Piperidinobutiophenone (hydrochloride), Purity: ≥98%, CAS: 92728-82-0, 5 mg
SDP
Catalog No. 502894334
Encompass_Preferred
An analytical reference standard categorized as a cathinone; intended for research and forensic applications
Catalog No. 50-289-4334 Supplier Cayman Chemical Supplier No. 90015135 MG
An analytical reference standard categorized as a cathinone; intended for research and forensic applications