Learn More
LGC GENOMICS LLC (Lucigen) Poly(A) Polymerase Tailing Kit, 50 RxnsAdd poly(A) tails to the 3' ends of RNA molecules, including in vitro transcribed RNAs, with this robust kit.
Catalog No. NC9401302
Encompass
The Poly(A) Polymerase Tailing Kit provides the enzyme and other reagents for quickly and easily adding a poly(A) tail to the 3 end of any RNA. Poly(A) Polymerase is encoded by an E. coli gene that has been cloned into a plasmid and overexpressed in an E. coli strain.
This item is not returnable. View return policy
Catalog No. NC9401302 Supplier LGC GENOMICS LLC Supplier No. PAP5104H
The Poly(A) Polymerase Tailing Kit provides the enzyme and other reagents for quickly and easily adding a poly(A) tail to the 3 end of any RNA. Poly(A) Polymerase is encoded by an E. coli gene that has been cloned into a plasmid and overexpressed in an E. coli strain.