Learn More
Cayman Chemical α-Propylaminopentiophenone (hydrochloride), Purity: ≥98%, CAS: 18268-15-0, 5 mg
SDP
Catalog No. 502966604
Encompass_Preferred
A substituted cathinone that has propyl groups both at the α position of the cathinone structure and at the amine
Catalog No. 50-296-6604 Supplier Cayman Chemical Supplier No. 170855 MG
A substituted cathinone that has propyl groups both at the α position of the cathinone structure and at the amine