Learn More
Cayman Chemical α-Pyrrolidinononanophenone (hydrochloride), Purity: ≥98%, CAS: 2748414-29-9, 50 mg
SDP
Catalog No. 502965381
Encompass_Preferred
A novel α-PVP analog that features a side chain extended in length by four methylene groups; intended for forensic and research applications
Catalog No. 50-296-5381 Supplier Cayman Chemical Supplier No. 1713850 MG
A novel α-PVP analog that features a side chain extended in length by four methylene groups; intended for forensic and research applications