Learn More
Cayman Chemical a-Pyrrolidinopentiophenone me
SDP
Catalog No. 502952832
Encompass_Preferred
An analog of α-PVP in which the keto group is reduced; intended for forensic and research purposes
Catalog No. 50-295-2832 Supplier Cayman Chemical Supplier No. 140935 MG
An analog of α-PVP in which the keto group is reduced; intended for forensic and research purposes