Learn More
Eurofins Discoverx A-Raf, active, 250 µg. (14-956M)
Supplier:  Eurofins Discoverx 14956M
Encompass
N-terminal GST-tagged recombinant, human A-Raf amino acids 273-end containing the mutations Y301D and Y302D. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9481