Learn More
MSC Value Collection Anti-Static Poly Bag, 12 x 12 in, 500/Pk.
Catalog No. 501920668
Encompass
This anti-static poly bag is designed to protect electronic components from static buildup, dirt, dust, and moisture.
  • Protects electronic components
  • Prevents static buildup
  • Shields against dirt, dust, and moisture
  • Pink color
  • Features 4 mil thickness
  • Measures 12 inch overall width
  • Measures 12 inch overall length
  • Supplied in packs of 500
Catalog No. 50-192-0668 Supplier MSC Supplier No. 39494638
This anti-static poly bag is designed to protect electronic components from static buildup, dirt, dust, and moisture.
  • Protects electronic components
  • Prevents static buildup
  • Shields against dirt, dust, and moisture
  • Pink color
  • Features 4 mil thickness
  • Measures 12 inch overall width
  • Measures 12 inch overall length
  • Supplied in packs of 500