Learn More
Apexbio Technology LLC A-SANTONIN-500mg
SDP
Catalog No. 5000897612
Encompass_Preferred
-santonin (CAS No 481-06-1) is a sesquiterpene lactone commonly studied in the field of biomedical research for its bioactive properties It is derived from Artemisia species particularly Artemisia maritima -santonin is known to influence biochemical pathways by interacting with specific cellular targets including enzymes and receptors that modulate inflammation and apoptosis Its in vitro activity often falls within the nanomolar to low micromolar range indicating significant potency This compound is frequently used in cell models and animal studies to explore its potential as an anti-inflammatory and antiparasitic agent as well as in drug screening programs to identify novel therapeutic candidates Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97612 Supplier Apexbio Technology LLC Supplier No. C6563500
-santonin (CAS No 481-06-1) is a sesquiterpene lactone commonly studied in the field of biomedical research for its bioactive properties It is derived from Artemisia species particularly Artemisia maritima -santonin is known to influence biochemical pathways by interacting with specific cellular targets including enzymes and receptors that modulate inflammation and apoptosis Its in vitro activity often falls within the nanomolar to low micromolar range indicating significant potency This compound is frequently used in cell models and animal studies to explore its potential as an anti-inflammatory and antiparasitic agent as well as in drug screening programs to identify novel therapeutic candidates Experimental concentrations typically depend on the specific research design