Learn More
ABclonal Technology -Smooth Muscle Actin (ACTA2) Rabbit mAb 100 L
SDP
Supplier:  ABclonal Technology A17910100UL
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 1-100 of human α-Smooth Muscle Actin (ACTA2) (P62736).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IHC,IF•Swiss Prot: P62736•Gene ID:59•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. NC2079860