Learn More
R Biopharm Inc Enzytec™ Generic Starch Kit, Quantitative Starch Determination, UV-Method, 29 Kits
Catalog No. NC2080260
Encompass
The Enzytec™ Generic Starch Kit is designed for the quantitative determination of starch in foodstuffs and other materials using a reliable UV-method. This kit allows for approximately 32 tests, delivering precise results with a detection limit of 6 mg Starch/L. Its convenient storage conditions and optimal operating temperatures enhance usability, making it an excellent choice for food and feed analysis, distinguishing it from other enzymatic kits.
Catalog No. NC2080260 Supplier R Biopharm Inc Supplier No. E1268
The Enzytec™ Generic Starch Kit is designed for the quantitative determination of starch in foodstuffs and other materials using a reliable UV-method. This kit allows for approximately 32 tests, delivering precise results with a detection limit of 6 mg Starch/L. Its convenient storage conditions and optimal operating temperatures enhance usability, making it an excellent choice for food and feed analysis, distinguishing it from other enzymatic kits.