Learn More
Ace Glass, Inc. PTFE stir blade, 19x76mm, 10mm shafts
SDP
Catalog No. 509558247
Encompass_Preferred
19x76mm PTFE stir blade for 10mm O.D. button type stir shafts.
Catalog No. 50-955-8247 Supplier Ace Glass, Inc. Supplier No. 808511
19x76mm PTFE stir blade for 10mm O.D. button type stir shafts.