Learn More
Bachem Fmoc-L-azetidine-2-carboxylic acid
SDP
Supplier:  Bachem 4019052.5000
Encompass_Preferred
Fmoc-L-azetidine-2-carboxylic acid, 136552-06-2, C19H17NO4, Mr 323.35, 5g. Fmoc-Aze-OH. A suitable derivative for incorporating this strained proline analog in peptides to modify their conformation. Its influence on the stability of the collagen helix has been studied by Zagari et al. Fmoc-Aze-OH was employed in the solid-phase synthesis of potent cyclic GnRH analogs.
Catalog No. 50-194-7133