Learn More
Metrohm USA A SUPP 5 ELUENT 20X CONC. 1L
SDP
Catalog No. NC1536090 Shop All Metrohm USA Products
Encompass
  • Designed for use with Metrosep A Supp 5 columns
  • Facilitates separation and detection of standard anions and carboxylic acids
  • 20X concentrated solution to be diluted to 1 L for working eluent
  • Compatible with automated IC systems
  • Streamlines laboratory workflows
  • Enhances analytical reproducibility
  • Precise formulation ensures optimal chromatographic performance
  • Applicable in water, environmental, pharmaceutical, and food analysis
  • Liquid form
  • Soluble in water
  • Odorless
  • Made in the United States
  • Suitable for inline eluent generators
Catalog No. NC1536090 Supplier Metrohm USA Supplier No. ERAIC1101
  • Designed for use with Metrosep A Supp 5 columns
  • Facilitates separation and detection of standard anions and carboxylic acids
  • 20X concentrated solution to be diluted to 1 L for working eluent
  • Compatible with automated IC systems
  • Streamlines laboratory workflows
  • Enhances analytical reproducibility
  • Precise formulation ensures optimal chromatographic performance
  • Applicable in water, environmental, pharmaceutical, and food analysis
  • Liquid form
  • Soluble in water
  • Odorless
  • Made in the United States
  • Suitable for inline eluent generators