Learn More
Medchemexpress LLC A synthetic superoxide dismutase and catalase mimetic | 81065-76-1 | 98.3% | 416.74 | C18H18ClMnN2O4 | 50 MG
SDP
Catalog No. 5000212963
Encompass_Preferred
EUK-134 is a synthetic small-molecule antioxidant with combined superoxide dismutase and catalase mimetic activity. It is supplied as a high-purity solid for research use in cell and animal models to investigate oxidative stress, mitochondrial protection, and related signaling pathways.
  • Catalytically scavenges reactive oxygen species (ROS).
  • Exhibits superoxide dismutase and catalase mimetic activity.
  • Used in vitro and in vivo to study oxidative stress and mitochondrial function.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as 50 mg solid for laboratory use.
Catalog No. 50-002-12963 Supplier Medchemexpress LLC Supplier No. HY10059450MG
EUK-134 is a synthetic small-molecule antioxidant with combined superoxide dismutase and catalase mimetic activity. It is supplied as a high-purity solid for research use in cell and animal models to investigate oxidative stress, mitochondrial protection, and related signaling pathways.
  • Catalytically scavenges reactive oxygen species (ROS).
  • Exhibits superoxide dismutase and catalase mimetic activity.
  • Used in vitro and in vivo to study oxidative stress and mitochondrial function.
  • High purity suitable for biochemical and cellular assays.
  • Supplied as 50 mg solid for laboratory use.