Learn More
Montana Molecular α-synuclein A53T BacMam - 5mL
SDP
Supplier:  Montana Molecular N0410N005
Encompass_Preferred
α-synuclein with the A53T mutation packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. Wild-type or versions with A30P, E46K, H50Q, and G51D mutations are also available. Co-express with Montana Molecular sensors for ER stress, cAMP, calcium, and more. This kit includes the α-synuclein A53T BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600438