Learn More
Montana Molecular α-synuclein G51D BacMam - 5mL
SDP
Supplier:  Montana Molecular N0450N005
Encompass_Preferred
α-synuclein with the G51D mutation packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. Wild-type or versions with A53T, A30P, E46K, and H50Q mutations are also available. Co-express with Montana Molecular sensors for ER stress, cAMP, calcium, and more. This kit includes the α-synuclein G51D BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600450