Learn More
Enzo Life Sciences alpha-Synuclein (human) monoclonal antibody (LB509) (0.5ml)
SDP
Catalog No. 502012769
Encompass_Preferred
Clone: LB509. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 115-122 of human α-synuclein. Species reactivity: Human. Applications: ELISA, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2769 Supplier Enzo Life Sciences Supplier No. ENZABS2180500
Clone: LB509. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 115-122 of human α-synuclein. Species reactivity: Human. Applications: ELISA, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.