Learn More
Enzo Life Sciences alpha-Synuclein (human) monoclonal antibody (LB509) (200µl)
SDP
Supplier:  Enzo Life Sciences ENZABS2180200
Encompass_Preferred
Clone: LB509. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to aa 115-122 of human α-synuclein. Species reactivity: Human. Applications: ELISA, IHC, WB. Purity: Purified. Formulation: Liquid. In PBS. Does not contain any preservatives or carrier proteins.
Catalog No. 50-201-2768