Learn More
Enzo Life Sciences alpha-Synuclein (human) monoclonal antibody (15G7) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX804258L001
Encompass_Preferred
Clone: 15G7. Isotype: IgG2a. Immunogen: Synthetic peptide corresponding to aa 116-131 of human α-synuclein. Species reactivity: Human. Applications: IHC (FS), IHC (PS), IP, WB. Formulation: Liquid. Hybridoma supernatant containing 0.05% sodium azide.
Catalog No. 50-200-9844