Learn More
Medchemexpress LLC Aβ/tau aggregation-IN-1 | 2252162-81-3 | 98.1% | C25H21IN2O | 10 MM 1 ML
SDP
Catalog No. 5000353740
Encompass_Preferred
Aβ/tau aggregation-IN-1 is a potent inhibitor of Aβ1-42 β-sheets formation and tau aggregation. With KD values of 160 μM for Aβ1-42 and 337 μM for tau, it effectively permeates the blood-brain barrier. Studies have shown its significant ability to reduce tau aggregation foci in HEK-293 T cells at a concentration of 1 μM for 24 hours. The compound achieves 84.7%-99.5% inhibition of Aβ1-42 aggregation and 71.2%-101.8% inhibition of tau aggregation by preventing β-sheet formation and through noncovalent interactions with both Aβ1-42 and tau.
  • Potent Aβ1-42 β-sheets formation inhibitor
  • Effective tau aggregation inhibitor
  • Permeates the blood-brain barrier
  • Reduces tau aggregation foci in HEK-293 T cells
  • Prevents β-sheet formation
  • Acts via noncovalent interactions with Aβ1-42 and tau
Catalog No. 50-003-53740 Supplier Medchemexpress LLC Supplier No. HY14166110MM/1ML
Aβ/tau aggregation-IN-1 is a potent inhibitor of Aβ1-42 β-sheets formation and tau aggregation. With KD values of 160 μM for Aβ1-42 and 337 μM for tau, it effectively permeates the blood-brain barrier. Studies have shown its significant ability to reduce tau aggregation foci in HEK-293 T cells at a concentration of 1 μM for 24 hours. The compound achieves 84.7%-99.5% inhibition of Aβ1-42 aggregation and 71.2%-101.8% inhibition of tau aggregation by preventing β-sheet formation and through noncovalent interactions with both Aβ1-42 and tau.
  • Potent Aβ1-42 β-sheets formation inhibitor
  • Effective tau aggregation inhibitor
  • Permeates the blood-brain barrier
  • Reduces tau aggregation foci in HEK-293 T cells
  • Prevents β-sheet formation
  • Acts via noncovalent interactions with Aβ1-42 and tau