Learn More
Medchemexpress LLC Aβ/tau aggregation-IN-3 | 1210750-32-5 | C23H22N4O3 | 5 MG

Supplier: Medchemexpress LLC HY1534305MG
Aβ/tau aggregation-IN-3 is a potent amyloid protein aggregation inhibitor with an IC50 of 0.85 μM by Aβ-Thioflavin T (Aβ-ThT) functional aggregation assay. It has anti-amyloid activity.
- Target: Amyloid-β
- Pathway: Neuronal Signaling
- Solvent & Solubility (In Vitro): DMSO: 50 mg/mL (124.24 mM; Need ultrasonic)
- Storage: 4°C, sealed, away from moisture and light
- Storage in solvent: -80°C, 6 months; -20°C, 1 month (sealed, away from moisture and light)
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.