Learn More
Cayman Chemical α-Terpinene, Purity: ≥95%, CAS: 99-86-5, 500 mg
SDP
Catalog No. 502986686
Encompass_Preferred
A terpenoid with diverse biological activities; induces 99% and 46% mortality of male and female T. putrescentiae larvae, respectively, at 32.3 µl/L via inhalation; increases longevity when used alone and decreases mortality when used in combination with diminazene aceturate in mice infected with T. evansi; scavenges DPPH radicals at 10 mM
Catalog No. 50-298-6686 Supplier Cayman Chemical Supplier No. 23178500 MG
A terpenoid with diverse biological activities; induces 99% and 46% mortality of male and female T. putrescentiae larvae, respectively, at 32.3 µl/L via inhalation; increases longevity when used alone and decreases mortality when used in combination with diminazene aceturate in mice infected with T. evansi; scavenges DPPH radicals at 10 mM