Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC A-terpinene | 99-86-5 | MFCD00001534 | 136.23 g/mol | C10H16 | 500 MG
SDP

Catalog No. 5000300946
Encompass_Preferred

α-Terpinene is a naturally occurring monoterpene (CAS 99-86-5) widely used as a research reagent and reference standard. It is found in essential oils, has reported antioxidant and antifungal activity, and has been investigated for antiparasitic effects in research studies.

  • Monoterpene occurring in essential oils.
  • Chemical formula C10H16.
  • Molecular weight 136.23 g/mol.
  • Reported antioxidant and antifungal activity.
  • Used as a research reagent and reference standard.
  • Typical storage -20°C as supplied by manufacturer.
  • Available in small pack sizes, including 500 mg.

Catalog No. 50-003-00946 Supplier Medchemexpress LLC Supplier No. HYW020182500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

α-Terpinene is a naturally occurring monoterpene (CAS 99-86-5) widely used as a research reagent and reference standard. It is found in essential oils, has reported antioxidant and antifungal activity, and has been investigated for antiparasitic effects in research studies.

  • Monoterpene occurring in essential oils.
  • Chemical formula C10H16.
  • Molecular weight 136.23 g/mol.
  • Reported antioxidant and antifungal activity.
  • Used as a research reagent and reference standard.
  • Typical storage -20°C as supplied by manufacturer.
  • Available in small pack sizes, including 500 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.