Learn More
Selleck Chemical LLC a-Terpineol
SDP
Catalog No. 5000003516
Encompass_Preferred
-Terpineol a natural monoterpene is the major component of essential oils of several species of aromatic plants such as Origanium vulgare L and Ocimum canum Sims which are widely used for medicinal purposes It is widely used in the manufacturing of cosmetics soaps perfumes antiseptic agents and is considered one of the most frequently used fragrance compounds -TPN has anticholinergic action and can block the interaction between PGE2 and GM1 receptors and cholera toxin
Catalog No. 50-000-03516 Supplier Selleck Chemical LLC Supplier No. S559425MG
-Terpineol a natural monoterpene is the major component of essential oils of several species of aromatic plants such as Origanium vulgare L and Ocimum canum Sims which are widely used for medicinal purposes It is widely used in the manufacturing of cosmetics soaps perfumes antiseptic agents and is considered one of the most frequently used fragrance compounds -TPN has anticholinergic action and can block the interaction between PGE2 and GM1 receptors and cholera toxin