Learn More
Cayman Chemical (±)-α-Tocopherol, Purity: ≥95%, CAS: 10191-41-0, 25 g
SDP
Catalog No. 502936814
Encompass_Preferred
A biologically active form of vitamin E; inhibits brominated diphenyl ether-47-induced increases in ROS and PGE2 production in HTR-8/SVneo cells at 20 μM; inhibits lipid peroxidation in HepG2 cells (IC50 = 24.5 μM)
Catalog No. 50-293-6814 Supplier Cayman Chemical Supplier No. 2598525 G
A biologically active form of vitamin E; inhibits brominated diphenyl ether-47-induced increases in ROS and PGE2 production in HTR-8/SVneo cells at 20 μM; inhibits lipid peroxidation in HepG2 cells (IC50 = 24.5 μM)