Learn More
Cayman Chemical (+)-α-Tocopherol Acetate, Purity: ≥95%, CAS: 58-95-7, 10 g
SDP
Catalog No. 502950342
Encompass_Preferred
An acetate form of (+)-α-tocopherol; reduces liver necrosis and increases survival in male rats in a model of CCl4-induced hepatotoxicity at 450 mg/kg three times at 72, 48, and 24 hours before CCl4 administration; decreases abdominal fat percentage and breast muscle MDA levels and increases breast muscle total antioxidant capacity, total SOD activity, and GPX activity in broiler chickens at 20 mg/kg in the diet
Catalog No. 50-295-0342 Supplier Cayman Chemical Supplier No. 3179510 G
An acetate form of (+)-α-tocopherol; reduces liver necrosis and increases survival in male rats in a model of CCl4-induced hepatotoxicity at 450 mg/kg three times at 72, 48, and 24 hours before CCl4 administration; decreases abdominal fat percentage and breast muscle MDA levels and increases breast muscle total antioxidant capacity, total SOD activity, and GPX activity in broiler chickens at 20 mg/kg in the diet