Learn More
Cayman Chemical (±)-α-Tocopherol Acetate-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2742364-46-9, 1 mg
SDP
Catalog No. 502989964
Encompass_Preferred
An internal standard for the quantification of (±)-α-tocopherol acetate by GC- or LC-MS
Catalog No. 50-298-9964 Supplier Cayman Chemical Supplier No. 300811 MG
An internal standard for the quantification of (±)-α-tocopherol acetate by GC- or LC-MS