Learn More
Apexbio Technology LLC A-TOCOPHEROL D6-1mg
SDP
Catalog No. 5000897647
Encompass_Preferred
-Tocopherol-d6 (CAS No 113892-08-3) is a deuterated form of vitamin E primarily used in biomedical research to study the antioxidant properties of tocopherols Originating from synthetic processes this compound plays a crucial role in research focused on oxidative stress lipid peroxidation and associated cellular pathways Its mechanism of action involves scavenging free radicals and modulating enzymatic activity related to reactive oxygen species In vitro studies typically report -Tocopherol-d6 activity in nanomolar to low micromolar ranges making it suitable for exploring cellular antioxidative responses Common applications include use in cell models such as hepatocytes or neural cells and in animal studies assessing oxidative damage and protective mechanisms It is also employed in drug screening to evaluate the efficacy of antioxidant therapies Experimental concentrations typically depend on the specific research design
Catalog No. 50-008-97647 Supplier Apexbio Technology LLC Supplier No. SL10021
Add to Cart
Add to Cart
-Tocopherol-d6 (CAS No 113892-08-3) is a deuterated form of vitamin E primarily used in biomedical research to study the antioxidant properties of tocopherols Originating from synthetic processes this compound plays a crucial role in research focused on oxidative stress lipid peroxidation and associated cellular pathways Its mechanism of action involves scavenging free radicals and modulating enzymatic activity related to reactive oxygen species In vitro studies typically report -Tocopherol-d6 activity in nanomolar to low micromolar ranges making it suitable for exploring cellular antioxidative responses Common applications include use in cell models such as hepatocytes or neural cells and in animal studies assessing oxidative damage and protective mechanisms It is also employed in drug screening to evaluate the efficacy of antioxidant therapies Experimental concentrations typically depend on the specific research design