Learn More
Medchemexpress LLC a-Tocopherol phospha 5mg
SDP
Catalog No. 5000204902
Encompass_Preferred
-Tocopherol phosphate disodium is an antioxidant that protects against long-wave UVA1 induced cell death and scavenge UVA1 induced ROS in a skin cell model -Tocopherol phosphate disodium exhibits angiogenesis-promoting activity[1][2]
Catalog No. 50-002-04902 Supplier Medchemexpress LLC Supplier No. HYW1275305MG
-Tocopherol phosphate disodium is an antioxidant that protects against long-wave UVA1 induced cell death and scavenge UVA1 induced ROS in a skin cell model -Tocopherol phosphate disodium exhibits angiogenesis-promoting activity[1][2]