Learn More
Cayman Chemical a-TruxIlIc AcId 50mg
SDP
Catalog No. 502972481
Encompass_Preferred
Can be formed by the dimerization of two molecules of α-trans-cinnamic acid; significantly blocks inflammatory pain while having little effect on neurogenic pain, as indicated by the formalin test in mice
Catalog No. 50-297-2481 Supplier Cayman Chemical Supplier No. 1486550 MG
Can be formed by the dimerization of two molecules of α-trans-cinnamic acid; significantly blocks inflammatory pain while having little effect on neurogenic pain, as indicated by the formalin test in mice