Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC a-Truxillic acid 100mg | 490-20-0 | 100 MG
SDP

Catalog No. 5000228286
Encompass_Preferred

α-Truxillic acid is a cyclobutane dicarboxylic acid formed by the dimerization of α-trans-cinnamic acid. It is supplied as a white to off-white solid for research use and has reported anti-inflammatory activity.

  • High purity: 99.9%.
  • White to off-white solid form.
  • Molecular formula C18H16O4.
  • Molecular weight 296.32 g/mol.
  • Available as a 100 MG package.

Catalog No. 50-002-28286 Supplier Medchemexpress LLC Supplier No. HY114771100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

α-Truxillic acid is a cyclobutane dicarboxylic acid formed by the dimerization of α-trans-cinnamic acid. It is supplied as a white to off-white solid for research use and has reported anti-inflammatory activity.

  • High purity: 99.9%.
  • White to off-white solid form.
  • Molecular formula C18H16O4.
  • Molecular weight 296.32 g/mol.
  • Available as a 100 MG package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.