Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC a-Truxillic acid 10mg | 490-20-0 | 10 MG
SDP

Catalog No. 5000226372
Encompass_Preferred

α-Truxillic acid (CAS 490-20-0) is the cyclobutane dimer of α-trans-cinnamic acid formed by [2+2] photodimerization and is reported to have anti-inflammatory activity. It is supplied as a white to off-white solid for research use; analytical and handling data (purity, solubility, and storage recommendations) are provided by the supplier.

  • High purity (99.93%).
  • Molecular formula C18H16O4; molecular weight 296.32 g/mol.
  • Appearance: white to off-white solid.
  • Soluble in DMSO (116.67 mg/mL); ultrasonic assistance recommended.
  • Powder storage: -20°C, stable for up to 3 years.
  • Available pack sizes include 10 mg, 25 mg, 50 mg, and 100 mg.
  • For research use only; not for patient use.

Catalog No. 50-002-26372 Supplier Medchemexpress LLC Supplier No. HY11477110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

α-Truxillic acid (CAS 490-20-0) is the cyclobutane dimer of α-trans-cinnamic acid formed by [2+2] photodimerization and is reported to have anti-inflammatory activity. It is supplied as a white to off-white solid for research use; analytical and handling data (purity, solubility, and storage recommendations) are provided by the supplier.

  • High purity (99.93%).
  • Molecular formula C18H16O4; molecular weight 296.32 g/mol.
  • Appearance: white to off-white solid.
  • Soluble in DMSO (116.67 mg/mL); ultrasonic assistance recommended.
  • Powder storage: -20°C, stable for up to 3 years.
  • Available pack sizes include 10 mg, 25 mg, 50 mg, and 100 mg.
  • For research use only; not for patient use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.