Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC a-Truxillic acid 10mM 1mL | 490-20-0 | 1 ML
SDP

Catalog No. 5000228995
Encompass_Preferred

Alpha-truxillic acid is the cyclobutane dimer of alpha-trans-cinnamic acid; it is reported to have anti-inflammatory activity and is supplied for research use as a high-purity solid and as a ready-to-use 10 mM solution in DMSO.

  • Reported anti-inflammatory activity
  • High purity (99.93%)
  • Molecular weight 296.32 g/mol
  • Available as solid powder and 10 mM solution in DMSO
  • Recommended storage: powder -20°C; in solvent -80°C (up to 6 months)

Catalog No. 50-002-28995 Supplier Medchemexpress LLC Supplier No. HY11477110MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Alpha-truxillic acid is the cyclobutane dimer of alpha-trans-cinnamic acid; it is reported to have anti-inflammatory activity and is supplied for research use as a high-purity solid and as a ready-to-use 10 mM solution in DMSO.

  • Reported anti-inflammatory activity
  • High purity (99.93%)
  • Molecular weight 296.32 g/mol
  • Available as solid powder and 10 mM solution in DMSO
  • Recommended storage: powder -20°C; in solvent -80°C (up to 6 months)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.