Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC a-Truxillic acid 25mg | 490-20-0 | 25 MG
SDP

Catalog No. 5000229292
Encompass_Preferred

α-Truxillic acid is the photodimerization product of two α-trans-cinnamic acid molecules. It is used as a research compound and drug intermediate and has reported anti-inflammatory activity. The compound has molecular formula C18H16O4, molecular weight 296.32 g/mol, and CAS 490-20-0.

  • Photodimer of α-trans-cinnamic acid
  • Reported anti-inflammatory activity
  • Used as a research compound and drug intermediate
  • Molecular formula C18H16O4
  • Molecular weight 296.32 g/mol
  • Commonly supplied in small sample sizes (25 mg)

Catalog No. 50-002-29292 Supplier Medchemexpress LLC Supplier No. HY11477125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

α-Truxillic acid is the photodimerization product of two α-trans-cinnamic acid molecules. It is used as a research compound and drug intermediate and has reported anti-inflammatory activity. The compound has molecular formula C18H16O4, molecular weight 296.32 g/mol, and CAS 490-20-0.

  • Photodimer of α-trans-cinnamic acid
  • Reported anti-inflammatory activity
  • Used as a research compound and drug intermediate
  • Molecular formula C18H16O4
  • Molecular weight 296.32 g/mol
  • Commonly supplied in small sample sizes (25 mg)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.