Learn More
Medchemexpress LLC a-Truxillic acid | 50MG
SDP
Catalog No. 5000227245
Encompass_Preferred
-Truxillic acid is form by the dimerization of two molecules of -trans-cinnamic acid with anti-inflammatory activities[1]
Catalog No. 50-002-27245 Supplier Medchemexpress LLC Supplier No. HY11477150MG
-Truxillic acid is form by the dimerization of two molecules of -trans-cinnamic acid with anti-inflammatory activities[1]