Learn More
Enzo Life Sciences alpha-Tubulin (K(Ac)40) polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA4520100
Encompass_Preferred
Immunogen: Synthetic acetylated peptide (K(Ac)40) corresponding to aa 35-45 (Q35MPSD-K(Ac)-TIGGG45) of human α-tubulin. Source: Purified from rabbit serum. Species reactivity: Zebrafish, Xenopus, Rodent, Primate, Chicken. Applications: WB. Formulation: Liquid. In PBS.
Catalog No. 50-201-1827