Learn More
AdipoGen anti-alpha-Tubulin, mAb (rec.) (F2C) (ATTO 488) (100 µg)
SDP
Supplier:  AdipoGen AG27B0005TDC100
Encompass_Preferred
Recombinant Antibody. Recognizes mouse, bovine and human alpha-tubulin. Species cross-reactivity: Bovine, Human, Mouse. Clone: F2C. Isotype: Human IgG2lambda. Applications: ICC. Host: Purified from HEK 293 cell culture supernatant. Liquid. In PBS containing 0.02% sodium azide. Tubulin is the major building block of microtubules. This intracellular cylindrical filamentous structure is present in almost all eukaryotic cells. Microtubules function as structural and mobile elements in mitosis, intracellular transport, flagellar movement, and the cytoskeleton.
Catalog No. 50-205-3023