Learn More
Grainger A/V Splitter HDMI M/Fx2
Catalog No. 502752479
Encompass_Preferred
The Monoprice 1x2 HDMI Splitter is a device that allows you to connect one HDMI source to two HDMI displays or receivers. It supports resolutions up to 4K@30Hz and has a bandwidth of 10.2 Gbps. The splitter has one HDMI input and two HDMI outputs. It is powered via a 5V micro USB port and comes with a 5V/1A power adapter. The splitter supports HDMI 1.4 and HDCP 1.4 copy protection. It has a compact and portable design in a plastic enclosure measuring just 3.5 x 1.4 x 0.6 inches. The Monoprice 1x2 HDMI Splitter provides an easy and affordable way to send an HDMI signal to two displays at the same time.
Catalog No. 50-275-2479 Supplier Grainger Supplier No. 14C186
The Monoprice 1x2 HDMI Splitter is a device that allows you to connect one HDMI source to two HDMI displays or receivers. It supports resolutions up to 4K@30Hz and has a bandwidth of 10.2 Gbps. The splitter has one HDMI input and two HDMI outputs. It is powered via a 5V micro USB port and comes with a 5V/1A power adapter. The splitter supports HDMI 1.4 and HDCP 1.4 copy protection. It has a compact and portable design in a plastic enclosure measuring just 3.5 x 1.4 x 0.6 inches. The Monoprice 1x2 HDMI Splitter provides an easy and affordable way to send an HDMI signal to two displays at the same time.