Learn More
Grainger A/V WallPlate HDMI w/Ext 2P White
Supplier:  Grainger 14J384
Encompass_Preferred
The Monoprice Audio Video Wall Plate is a single gang wall plate with two female RCA jacks and one female HDMI port. It is designed for in-wall installations to provide connection points for component video, composite video, stereo audio and digital HDMI signals. The connectors are gold plated for optimal signal transfer. The dimensions are 4.5 x 2.75 x 1.4 inches. It comes in white color and has mounting screws included. The wall plate provides a clean, integrated solution for connecting A/V equipment through walls.
Catalog No. 50-275-1267