Learn More
Bachem Pyr-OH
SDP
Supplier:  Bachem 4026127.0500
Encompass_Preferred
Pyr-OH, 98-79-3, C5H7NO3, Mr 129.12, 500g. L-Pyroglutamic acid. A versatile chiral synthon. Panday et al. reviewed its use in asymmetric synthesis.
Catalog No. 50-194-7298