Learn More
ABclonal Technology FOXO3A Rabbit pAb
SDP
Catalog No. 5000137504
Encompass_Preferred
Enables DNA binding activity, DNA-binding transcription factor activity, RNA polymerase II-specific, and mitochondrial transcription factor activity. Involved in several processes, including mitochondrial transcription, positive regulation of muscle atrophy, and positive regulation of pri-miRNA transcription by RNA polymerase II. Acts upstream of or within several processes, including extrinsic apoptotic signaling pathway in absence of ligand, female gonad development, and neuronal stem cell population maintenance. Located in cytosol, mitochondrial outer membrane, and nucleus. Part of protein-containing complex. Colocalizes with mitochondrial matrix. Is expressed in several structures, including alimentary system, brain, early embryo, genitourinary system, and hemolymphoid system. Used to study dermoid cyst of ovary. Orthologous to human FOXO3 (forkhead box O3).
Catalog No. 50-001-37504 Supplier ABclonal Technology Supplier No. A01021000UL
Enables DNA binding activity, DNA-binding transcription factor activity, RNA polymerase II-specific, and mitochondrial transcription factor activity. Involved in several processes, including mitochondrial transcription, positive regulation of muscle atrophy, and positive regulation of pri-miRNA transcription by RNA polymerase II. Acts upstream of or within several processes, including extrinsic apoptotic signaling pathway in absence of ligand, female gonad development, and neuronal stem cell population maintenance. Located in cytosol, mitochondrial outer membrane, and nucleus. Part of protein-containing complex. Colocalizes with mitochondrial matrix. Is expressed in several structures, including alimentary system, brain, early embryo, genitourinary system, and hemolymphoid system. Used to study dermoid cyst of ovary. Orthologous to human FOXO3 (forkhead box O3).