Learn More
ABclonal Technology DOK4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A0226500UL
Encompass_Preferred
Predicted to be involved in positive regulation of MAPK cascade and transmembrane receptor protein tyrosine kinase signaling pathway. Predicted to act upstream of or within nervous system development. Predicted to be located in cytosol. Predicted to be active in cytoplasm.
Catalog No. 50-001-38786