Learn More
ABclonal Technology c-Fos Rabbit pAb
SDP
Catalog No. 5000135760
Encompass_Preferred
The Fos gene family consists of 4 members FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death.
Catalog No. 50-001-35760 Supplier ABclonal Technology Supplier No. A0236500UL
The Fos gene family consists of 4 members FOS, FOSB, FOSL1, and FOSL2. These genes encode leucine zipper proteins that can dimerize with proteins of the JUN family, thereby forming the transcription factor complex AP-1. As such, the FOS proteins have been implicated as regulators of cell proliferation, differentiation, and transformation. In some cases, expression of the FOS gene has also been associated with apoptotic cell death.