Learn More
ABclonal Technology MTM1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A02551000UL
Encompass_Preferred
This gene encodes a dual-specificity phosphatase that acts on both phosphotyrosine and phosphoserine. It is required for muscle cell differentiation and mutations in this gene have been identified as being responsible for X-linked myotubular myopathy.
Catalog No. 50-001-34983