Learn More
ABclonal Technology MTM1 Rabbit pAb
SDP
Catalog No. 5000138716
Encompass_Preferred
This gene encodes a dual-specificity phosphatase that acts on both phosphotyrosine and phosphoserine. It is required for muscle cell differentiation and mutations in this gene have been identified as being responsible for X-linked myotubular myopathy.
Catalog No. 50-001-38716 Supplier ABclonal Technology Supplier No. A0255500UL
This gene encodes a dual-specificity phosphatase that acts on both phosphotyrosine and phosphoserine. It is required for muscle cell differentiation and mutations in this gene have been identified as being responsible for X-linked myotubular myopathy.