Learn More
ABclonal Technology CRM1/XPO1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A02991000UL
Encompass_Preferred
This cell-cycle-regulated gene encodes a protein that mediates leucine-rich nuclear export signal (NES)-dependent protein transport. The protein specifically inhibits the nuclear export of Rev and U snRNAs. It is involved in the control of several cellular processes by controlling the localization of cyclin B, MPAK, and MAPKAP kinase 2. This protein also regulates NFAT and AP-1.
Catalog No. 50-001-37495