Learn More
ABclonal Technology MTHFR Rabbit pAb
SDP
Supplier:  ABclonal Technology A03051000UL
Encompass_Preferred
The protein encoded by this gene catalyzes the conversion of 5,10-methylenetetrahydrofolate to 5-methyltetrahydrofolate, a co-substrate for homocysteine remethylation to methionine. Genetic variation in this gene influences susceptibility to occlusive vascular disease, neural tube defects, colon cancer and acute leukemia, and mutations in this gene are associated with methylenetetrahydrofolate reductase deficiency.
Catalog No. 50-001-38695