Learn More
ABclonal Technology Hsp90α Rabbit pAb
SDP
Supplier:  ABclonal Technology A03651000UL
Encompass_Preferred
The protein encoded by this gene is an inducible molecular chaperone that functions as a homodimer. The encoded protein aids in the proper folding of specific target proteins by use of an ATPase activity that is modulated by co-chaperones. Two transcript variants encoding different isoforms have been found for this gene.
Catalog No. 50-001-35069